cis-tert-Butyl-3-iodocyclobutylcarbamate

Supplier: Matrix Scientific

113218-1G
102891-680EA 1890.77 USD
102891-680
cis-tert-Butyl-3-iodocyclobutylcarbamate
cis-tert-Butyl-3-iodocyclobutylcarbamate

Hazard: Irritant

Formula: C₉H₁₆INO₂
MW: 297.14 g/mol
MDL Number: MFCD22415233
CAS Number: 1389264-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR