3,3-Difluorocyclobutanol

Supplier: Matrix Scientific

113212-1G
102891-674EA 1309.23 USD
102891-674
3,3-Difluorocyclobutanol
3,3-Difluorocyclobutanol

Hazard: Irritant

Formula: C₄H₆F₂O
MDL Number: MFCD18791191
CAS Number: 637031-88-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR