5-Fluoro-2-thiophenemethamine

Supplier: Matrix Scientific

113209-1G
102891-666EA 2269.23 USD
102891-666
5-Fluoro-2-thiophenemethamine
5-Fluoro-2-thiophenemethamine

Hazard: Irritant

Formula: C₅H₆FNS
MDL Number: MFCD18324169
CAS Number: 214759-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR