Ethyl-trans-1-boc-4-hydroxypyrrolidine-3-carboxylate

Supplier: Matrix Scientific

113195-1G
102891-634EA 846.15 USD
102891-634
Ethyl-trans-1-boc-4-hydroxypyrrolidine-3-carboxylate
Ethyl-trans-1-boc-4-hydroxypyrrolidine-3-carboxylate

Hazard: Irritant

Formula: C₁₂H₂₁NO₅
MDL Number: MFCD23105897
CAS Number: 849935-83-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR