Methyl-4-methyl-5-nitropyridine-2-carboxylate

Supplier: Matrix Scientific

Synonyms: Methyl 4-methyl-5-nitropicolinate

113189-1G
102897-628EA 1309.23 USD
102897-628
Methyl-4-methyl-5-nitropyridine-2-carboxylate
Methyl-4-methyl-5-nitropyridine-2-carboxylate

Hazard: Irritant

Formula: C₈H₈N₂O₄
MW: 196.16 g/mol
MDL Number: MFCD22421724
CAS Number: 868551-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR