Methyl-1-methyl-4-nitro-1H-pyrazole-3-carboxylate

Supplier: Matrix Scientific

Synonyms: Methyl 1-methyl-4-nitro-1H-pyrazole-3-carboxylate

113187-5G
102897-626EA 376.92 USD
102897-626
Methyl-1-methyl-4-nitro-1H-pyrazole-3-carboxylate
Methyl-1-methyl-4-nitro-1H-pyrazole-3-carboxylate

Hazard: Irritant

Formula: C₆H₇N₃O₄
MW: 185.14 g/mol
MDL Number: MFCD19443825
CAS Number: 400877-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR