5-Amino-3-(difluoromethyl)-1H-pyrazole

Supplier: Matrix Scientific

Synonyms: 5-(Difluoromethyl)-1H-pyrazol-3-amine

113178-1G
102897-606EA 1309.23 USD
102897-606
5-Amino-3-(difluoromethyl)-1H-pyrazole
5-Amino-3-(difluoromethyl)-1H-pyrazole

Hazard: Irritant

Formula: C₄H₅F₂N₃
MW: 133.1 g/mol
MDL Number: MFCD23105881
CAS Number: 1284220-49-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR