3-(Boc-amino)oxetane-3-methanol

Supplier: Matrix Scientific

113173-1G
102891-622EA 2269.23 USD
102891-622
3-(Boc-amino)oxetane-3-methanol
3-(Boc-amino)oxetane-3-methanol

Hazard: Irritant

Formula: C₉H₁₇NO₄
MDL Number: MFCD22415250
CAS Number: 1363382-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR