(R)-Methyl-2-azetidinecarboxylate hydrochloride
Supplier: Matrix Scientific
113172-5G
113172-1G
102891-620EA
2000
USD
102891-620
102891-618
(R)-Methyl-2-azetidinecarboxylate hydrochloride
(R)-Methyl-2-azetidinecarboxylate hydrochloride
Hazard: Irritant
Formula:
C₅H₁₀ClNO₂ MW: 151.59 g/mol |
MDL Number:
MFCD22418536 CAS Number: 647854-63-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...