Ethyl-1-methylpyrazole-4-carboxylate

Supplier: Matrix Scientific

Synonyms: Ethyl 1-methyl-1H-pyrazole-4-carboxylate

113170-5G
102897-784EA 255.38 USD
102897-784
Ethyl-1-methylpyrazole-4-carboxylate
Ethyl-1-methylpyrazole-4-carboxylate

Hazard: Irritant

Formula: C₇H₁₀N₂O₂
MW: 154.17 g/mol
MDL Number: MFCD12180234
CAS Number: 85290-80-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR