Ethyl-4-hydroxypyrrolo[1,2-f][1,2,4]triazine-6-carboxylate

Supplier: Matrix Scientific

113169-1G
102897-780EA 1309.23 USD
102897-780
Ethyl-4-hydroxypyrrolo[1,2-f][1,2,4]triazine-6-carboxylate
Ethyl-4-hydroxypyrrolo[1,2-f][1,2,4]triazine-6-carboxylate

Hazard: Irritant

Formula: C₉H₉N₃O₃
MW: 207.19 g/mol
MDL Number: MFCD11976588
CAS Number: 869067-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR