5-Methyl-4-phenoxypyrrolo[2,1-f][1,2,4]triazine-6-carboxylic acid ethyl ester

Supplier: Matrix Scientific

113168-1G
102897-778EA 1309.23 USD
102897-778
5-Methyl-4-phenoxypyrrolo[2,1-f][1,2,4]triazine-6-carboxylic acid ethyl ester
5-Methyl-4-phenoxypyrrolo[2,1-f][1,2,4]triazine-6-carboxylic acid ethyl ester

Hazard: Irritant

Formula: C₁₆H₁₅N₃O₃
MW: 297.32 g/mol
MDL Number: MFCD22415213
CAS Number: 1363380-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR