trans-3-Amino-1-Cbz-4-hydroxypiperidine

Supplier: Matrix Scientific

113166-1G
102897-774EA 1687.69 USD
102897-774
trans-3-Amino-1-Cbz-4-hydroxypiperidine
trans-3-Amino-1-Cbz-4-hydroxypiperidine

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₃
MDL Number: MFCD22421662
CAS Number: 167832-26-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR