6-Iodo-3-methyl-1H-indazole

Supplier: Matrix Scientific

113165-1G
102897-772EA 1309.23 USD
102897-772
6-Iodo-3-methyl-1H-indazole
6-Iodo-3-methyl-1H-indazole

Hazard: Irritant

Formula: C₈H₇IN₂
MDL Number: MFCD11007870
CAS Number: 1082041-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR