6-Chloropyridazine-4-carboxylic acid

Supplier: Matrix Scientific

113161-1G
102897-764EA 176.92 USD
102897-764
6-Chloropyridazine-4-carboxylic acid
6-Chloropyridazine-4-carboxylic acid

Hazard: Irritant

Formula: C₅H₃ClN₂O₂
MDL Number: MFCD15146792
CAS Number: 1256794-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR