4-Chloroimidazo[2,1-f][1,2,4]triazine

Supplier: Matrix Scientific

113158-1G
102897-342EA 1890.77 USD
102897-342
4-Chloroimidazo[2,1-f][1,2,4]triazine
4-Chloroimidazo[2,1-f][1,2,4]triazine

Hazard: Irritant

Formula: C₅H₃ClN₄
MDL Number: MFCD19707604
CAS Number: 1206825-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR