8-Amino-3-bromoimidazo[1,2-a]pyrazine

Supplier: Matrix Scientific

Synonyms: 3-Bromoimidazo[1,2-a]pyrazin-8-amine

113154-1G
102891-566EA 1309.23 USD
102891-566
8-Amino-3-bromoimidazo[1,2-a]pyrazine
8-Amino-3-bromoimidazo[1,2-a]pyrazine

Hazard: Irritant

Formula: C₆H₅BrN₄
MW: 213.04 g/mol
MDL Number: MFCD09265736
CAS Number: 117718-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR