(2S,4S)-2-Methylpiperidin-4-ol hydrochloride
Supplier: Matrix Scientific
113145-1G
102891-550EA
2873.85
USD
102891-550
(2S,4S)-2-Methylpiperidin-4-ol hydrochloride
(2S,4S)-2-Methylpiperidin-4-ol hydrochloride
Hazard: Irritant
Formula:
C₆H₁₄ClNO |
MDL Number:
MFCD23105869 CAS Number: 103539-63-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...