2-Methylpiperidin-4-ol

Supplier: Matrix Scientific

113144-1G
102891-548EA 1018.46 USD
102891-548
2-Methylpiperidin-4-ol
2-Methylpiperidin-4-ol

Hazard: Irritant

Formula: C₆H₁₃NO
MDL Number: MFCD03307815
CAS Number: 344329-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR