1-Boc-3,3-difluoro-4,4-(dihydroxy)piperidine

Supplier: Matrix Scientific

Synonyms: 1-Boc-3,3-difluoro-4,4-dihydroxypiperidine3,3-Difluoro-4,4-dihydroxypiperidine-1-carboxylic acid tert-Butyl estertert-Butyl 3,3-Difluoro-4,4-dihydroxypiperidine-1-carboxylate1-(tert-Butoxycarbonyl)-3,3-difluoro-4,4-dihydroxypiperidine

113129-1G
102891-518EA 1600 USD
102891-518
1-Boc-3,3-difluoro-4,4-(dihydroxy)piperidine
1-Boc-3,3-difluoro-4,4-(dihydroxy)piperidine

Hazard: Irritant

Formula: C₁₀H₁₇F₂NO₄
MW: 253.25 g/mol
MDL Number: MFCD18072968
CAS Number: 1067914-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR