1-Boc-3-methylenepyrrolidine

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3-methylenepyrrolidine-1-carboxylatetert-Butyl 3-methylidenepyrrolidine-1-carboxylate

113118-5G 113118-1G
102897-728EA 2000 USD
102897-728 102897-726
1-Boc-3-methylenepyrrolidine
1-Boc-3-methylenepyrrolidine

Hazard: Irritant

Formula: C₁₀H₁₇NO₂
MW: 183.25 g/mol
MDL Number: MFCD12407903
CAS Number: 114214-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR