Ethyl-6-oxaspiro[2.5]octane-1-carboxylate

Supplier: Matrix Scientific

113109-1G
102891-500EA 1018.46 USD
102891-500
Ethyl-6-oxaspiro[2.5]octane-1-carboxylate
Ethyl-6-oxaspiro[2.5]octane-1-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₆O₃
MW: 184.24 g/mol
MDL Number: MFCD23105848
CAS Number: 909406-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR