2-Cbz-2,7-diazaspiro[4.4]nonane

Supplier: Matrix Scientific

113106-1G
102891-496EA 2001.54 USD
102891-496
2-Cbz-2,7-diazaspiro[4.4]nonane
2-Cbz-2,7-diazaspiro[4.4]nonane

Hazard: Irritant

Formula: C₁₅H₂₀N₂O₂
MDL Number: MFCD11223518
CAS Number: 1086394-74-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR