2-Amino-3-chloro-5-iodopyrazine

Supplier: Matrix Scientific

Synonyms: 3-Chloro-5-iodopyrazin-2-amine

113103-1G 113103-5G
102891-488EA 727.69 USD
102891-488 102891-490
2-Amino-3-chloro-5-iodopyrazine
2-Amino-3-chloro-5-iodopyrazine

Hazard: Irritant

Formula: C₄H₃ClIN₃
MW: 255.45 g/mol
MDL Number: MFCD14706048
CAS Number: 1252597-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR