Etravirine

Supplier: Matrix Scientific

Synonyms: 4-((6-Amino-5-bromo-2-((4-cyanophenyl)amino)pyrimidin-4-yl)oxy)-3,5-dimethylbenzonitrile

113100-1G
102891-482EA 1687.69 USD
102891-482
Etravirine
Etravirine

Hazard: Irritant

Formula: C₂₀H₁₅BrN₆O
MW: 435.29 g/mol
MDL Number: MFCD09837879
CAS Number: 269055-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR