4-Fluoro-tetrahydro-2H-pyran-4-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 4-Fluorotetrahydro-2H-pyran-4-carboxylic acid

113089-1G
102891-448EA 1978.46 USD
102891-448
4-Fluoro-tetrahydro-2H-pyran-4-carboxylic acid
4-Fluoro-tetrahydro-2H-pyran-4-carboxylic acid

Hazard: Irritant

Formula: C₆H₉FO₃
MW: 148.14 g/mol
MDL Number: MFCD12024536
CAS Number: 1150617-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR