4-Hydroxymethyl-4-fluorotetrahydro-2H-pyran

Supplier: Matrix Scientific

113088-1G
102891-446EA 1309.23 USD
102891-446
4-Hydroxymethyl-4-fluorotetrahydro-2H-pyran
4-Hydroxymethyl-4-fluorotetrahydro-2H-pyran

Hazard: Irritant

Formula: C₆H₁₁FO₂
MDL Number: MFCD18072006
CAS Number: 883442-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR