7-Cbz-1,7-diazaspiro[3.5]nonane

Supplier: Matrix Scientific

113072-1G
102897-682EA 2850.77 USD
102897-682
7-Cbz-1,7-diazaspiro[3.5]nonane
7-Cbz-1,7-diazaspiro[3.5]nonane

Hazard: Irritant

Formula: C₁₅H₂₀N₂O₂
MW: 260.34 g/mol
MDL Number: MFCD17016170
CAS Number: 1250999-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR