1-Boc-3-fluoropyrrolidine-3-methylamine

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3-(aminomethyl)-3-fluoropyrrolidine-1-carboxylate

113065-1G
102891-438EA 1744.62 USD
102891-438
1-Boc-3-fluoropyrrolidine-3-methylamine
1-Boc-3-fluoropyrrolidine-3-methylamine

Hazard: Irritant

Formula: C₁₀H₁₉FN₂O₂
MW: 218.27 g/mol
MDL Number: MFCD18375054
CAS Number: 1228765-06-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR