1-Boc-3-fluoropiperidine-3-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 1-(tert-Butoxycarbonyl)-3-fluoropiperidine-3-carboxylic acid

113063-1G
102891-434EA 1600 USD
102891-434
1-Boc-3-fluoropiperidine-3-carboxylic acid
1-Boc-3-fluoropiperidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₈FNO₄
MW: 247.27 g/mol
MDL Number: MFCD11847195
CAS Number: 934342-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR