Diisopropyl-3-hydroxycyclobutane-1,1-dicarboxylate
Supplier: Matrix Scientific
113053-1G
113053-5G
102897-598EA
933.85
USD
102897-598
102897-596
Diisopropyl-3-hydroxycyclobutane-1,1-dicarboxylate
Diisopropyl-3-hydroxycyclobutane-1,1-dicarboxylate
Hazard: Irritant
Formula:
C₁₂H₂₀O₅ MW: 244.29 g/mol |
MDL Number:
MFCD11869277 CAS Number: 869109-31-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...