1-Boc-3-ethynyl-3-hydroxyazetidine

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3-ethynyl-3-hydroxyazetidine-1-carboxylate

113034-1G
102891-408EA 1135.38 USD
102891-408
1-Boc-3-ethynyl-3-hydroxyazetidine
1-Boc-3-ethynyl-3-hydroxyazetidine

Hazard: Irritant

Formula: C₁₀H₁₅NO₃
MW: 197.24 g/mol
MDL Number: MFCD21324699
CAS Number: 1259034-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR