3-Cyclopropyl-3-hydroxyazetidine hydrochloride

Supplier: Matrix Scientific

113030-1G
102891-400EA 1106.15 USD
102891-400
3-Cyclopropyl-3-hydroxyazetidine hydrochloride
3-Cyclopropyl-3-hydroxyazetidine hydrochloride

Hazard: Irritant

Formula: C₆H₁₂ClNO
MDL Number: MFCD13248563
CAS Number: 848192-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR