1-Boc-3-(hydroxymethyl)-3-methylazetidine

Supplier: Matrix Scientific

Synonyms: 3-(Hydroxymethyl)-3-methylazetidine, N-BOC protected

113027-1G
102891-390EA 1053.85 USD
102891-390
1-Boc-3-(hydroxymethyl)-3-methylazetidine
1-Boc-3-(hydroxymethyl)-3-methylazetidine

Hazard: Irritant

Formula: C₁₀H₁₉NO₃
MW: 201.27 g/mol
MDL Number: MFCD20926177
CAS Number: 1363382-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR