1-Boc-3-fluoroazetidine-3-methanol

Supplier: Matrix Scientific

Synonyms: tert-Butyl 3-fluoro-3-(hydroxymethyl)azetidine-1-carboxylate

113026-1G
102891-388EA 2472.31 USD
102891-388
1-Boc-3-fluoroazetidine-3-methanol
1-Boc-3-fluoroazetidine-3-methanol

Hazard: Irritant

Formula: C₉H₁₆FNO₃
MW: 205.23 g/mol
MDL Number: MFCD17392630
CAS Number: 1126650-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR