Methyl-1-boc-3-fluoroazetidine-3-carboxylate

Supplier: Matrix Scientific

113025-1G
102891-386EA 2472.31 USD
102891-386
Methyl-1-boc-3-fluoroazetidine-3-carboxylate
Methyl-1-boc-3-fluoroazetidine-3-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₆FNO₄
MW: 233.24 g/mol
MDL Number: MFCD20922778
CAS Number: 1363382-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR