3-Boc-3,7-diazabicyclo[4.2.0]octane

Supplier: Matrix Scientific

113013-1G
102897-570EA 2269.23 USD
102897-570
3-Boc-3,7-diazabicyclo[4.2.0]octane
3-Boc-3,7-diazabicyclo[4.2.0]octane

Hazard: Irritant

Formula: C₁₁H₂₀N₂O₂
MDL Number: MFCD08234914
CAS Number: 885271-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR