1-Boc-2-methylazetidine-2-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 2-Methylazetidine-2-carboxylic acid, N-BOC protected

113010-1G
102891-370EA 1309.23 USD
102891-370
1-Boc-2-methylazetidine-2-carboxylic acid
1-Boc-2-methylazetidine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₁₇NO₄
MW: 215.25 g/mol
MDL Number: MFCD12406956
CAS Number: 449758-77-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR