3-Boc-3,9-diazabicyclo[4.2.1]nonane

Supplier: Matrix Scientific

113009-1G
102891-368EA 2181.54 USD
102891-368
3-Boc-3,9-diazabicyclo[4.2.1]nonane
3-Boc-3,9-diazabicyclo[4.2.1]nonane

Hazard: Irritant

Formula: C₁₂H₂₂N₂O₂
MDL Number: MFCD13196397
CAS Number: 286947-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR