3-Hydroxy-3-(trifluoromethyl)cyclobutanecarboxylic acid

Supplier: Matrix Scientific

113005-1G
102891-358EA 867.69 USD
102891-358
3-Hydroxy-3-(trifluoromethyl)cyclobutanecarboxylic acid
3-Hydroxy-3-(trifluoromethyl)cyclobutanecarboxylic acid

Hazard: Irritant

Formula: C₆H₇F₃O₃
MW: 184.12 g/mol
MDL Number: MFCD11870383
CAS Number: 1163729-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR