4-Amino-3-iodo-1-methyl-1H-pyrazolo[3,4-d]pyrimidine
Supplier: Matrix Scientific
Synonyms:
3-Iodo-1-methyl-1H-pyrazolo[3,4-d]pyrimidin-4-amine
112998-1G
112998-5G
102891-346EA
2000
USD
102891-346
102891-344
4-Amino-3-iodo-1-methyl-1H-pyrazolo[3,4-d]pyrimidine
4-Amino-3-iodo-1-methyl-1H-pyrazolo[3,4-d]pyrimidine
Hazard: Irritant
Formula:
C₆H₆IN₅ MW: 275.05 g/mol |
MDL Number:
MFCD22566154 CAS Number: 862729-12-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...