(2S,4S)-1-Boc-2-hydroxymethyl-4-methylpyrrolidine

Supplier: Matrix Scientific

112996-1G
102891-340EA 2850.77 USD
102891-340
(2S,4S)-1-Boc-2-hydroxymethyl-4-methylpyrrolidine
(2S,4S)-1-Boc-2-hydroxymethyl-4-methylpyrrolidine

Hazard: Irritant

Formula: C₁₁H₂₁NO₃
MDL Number: MFCD18071966
CAS Number: 540501-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR