1-Oxa-6-azaspiro[3.3]heptane oxalate

Supplier: Matrix Scientific

112995-1G
102891-338EA 2269.23 USD
102891-338
1-Oxa-6-azaspiro[3.3]heptane oxalate
1-Oxa-6-azaspiro[3.3]heptane oxalate

Hazard: Irritant

Formula: C₇H₁₁NO₅
MDL Number: MFCD17214340
CAS Number: 936947-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR