7-Boc-2-oxo-1,7-diazaspiro[3.5]nonane

Supplier: Matrix Scientific

112992-1G
102891-334EA 1500 USD
102891-334
7-Boc-2-oxo-1,7-diazaspiro[3.5]nonane
7-Boc-2-oxo-1,7-diazaspiro[3.5]nonane

Hazard: Irritant

Formula: C₁₂H₂₀N₂O₃
MW: 240.3 g/mol
MDL Number: MFCD13184428
CAS Number: 392331-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR