3-Bromo-4-fluorobenzenesulfonyl chloride ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Bromo-4-fluorobenzene-1-sulphonyl chloride
, 3-Bromo-4-fluorobenzene-1-sulfonyl chloride
087313-1G
087313-5G
102853-294EA
275.38
USD
102853-294
102853-292
3-Bromo-4-fluorobenzenesulfonyl chloride ≥97%
3-Bromo-4-fluorobenzenesulfonyl chloride
Hazard: Corrosive/Refrigerate
Formula:
C₆H₃BrClFO₂S MW: 273.51 g/mol |
MDL Number:
MFCD13185363 CAS Number: 631912-19-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...