tert-Butyl-2,6-diazaspiro[4.5]decane-6-carboxylate hydrochloride ≥95%

Supplier: Matrix Scientific

087300-500MG
102853-276EA 2396.92 USD
102853-276
tert-Butyl-2,6-diazaspiro[4.5]decane-6-carboxylate hydrochloride ≥95%
tert-Butyl-2,6-diazaspiro[4.5]decane-6-carboxylate hydrochloride

Hazard: Irritant

Formula: C₁₃H₂₅ClN₂O₂
MW: 276.81 g/mol
Melting Pt: 113…115 °C
MDL Number: MFCD22573520
CAS Number: 1407522-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR