Methyl-4-(2H-1,2,3,4-tetrazol-5-yl)benzoate ≥97%

Supplier: Matrix Scientific

087288-1G 087288-500MG
102853-226EA 666.15 USD
102853-226 102853-224
Methyl-4-(2H-1,2,3,4-tetrazol-5-yl)benzoate ≥97%
Methyl-4-(2H-1,2,3,4-tetrazol-5-yl)benzoate

Hazard: Irritant

Formula: C₉H₈N₄O₂
MDL Number: MFCD20441656
CAS Number: 82544-82-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR