Methyl-4-chloro-1-benzofuran-2-carboxylate ≥95%
Supplier: Matrix Scientific
087277-500MG
102853-182EA
529.23
USD
102853-182
Methyl-4-chloro-1-benzofuran-2-carboxylate ≥95%
Methyl-4-chloro-1-benzofuran-2-carboxylate
Hazard: Irritant
Formula:
C₁₀H₇ClO₃ Melting Pt: 90…92 °C |
MDL Number:
MFCD22573531 CAS Number: 1407521-97-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...