4-Iodo-1-acetyl-7-azaindole ≥97%
Supplier: Matrix Scientific
087261-5G
087261-500MG
087261-1G
102853-118EA
480
USD
102853-118
102853-116
102853-120
4-Iodo-1-acetyl-7-azaindole ≥97%
4-Iodo-1-acetyl-7-azaindole
Hazard: Irritant
Formula:
C₉H₇IN₂O Melting Pt: 138…139 °C |
MDL Number:
MFCD09880006 CAS Number: 443729-67-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...