2-[(2-Fluoro-6-nitrophenyl)sulfanyl]acetic acid ≥97%
Supplier: Matrix Scientific
087256-500MG
087256-1G
102853-098EA
749.23
USD
102853-098
102853-096
2-[(2-Fluoro-6-nitrophenyl)sulfanyl]acetic acid ≥97%
2-[(2-Fluoro-6-nitrophenyl)sulfanyl]acetic acid
Hazard: Irritant
Formula:
C₈H₆FNO₄S MW: 231.2 g/mol |
MDL Number:
MFCD22573544 CAS Number: 1247021-61-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...